Merck
  • Electrodialytic removal of nitrate from pineapple juice: effect on selected physicochemical properties, amino acids, and aroma components of the juice.
Electrodialytic removal of nitrate from pineapple juice: effect on selected physicochemical properties, amino acids, and aroma components of the juice.
Journal of food science (2015-04-02)
Yuwadee Ackarabanpojoue, Nathamol Chindapan, Tipaporn Yoovidhya, Sakamon Devahastin
ABSTRACT
This study aimed at investigating the effect of nitrate removal from pineapple juice by electrodialysis (ED) on selected properties of the ED-treated juice. Single-strength pineapple juice with reduced pulp content was treated by ED to reduce the nitrate concentration to 15, 10, or 5 ppm. After ED, the removed pulp was added to the ED-treated juice and its properties, including electrical conductivity, acidity, pH, total soluble solids (TSS), color, amino acids, and selected aroma compounds, were determined and compared with those of the untreated juice. ED could reduce the nitrate content of 1 L of pineapple juice from an initial value of 50 ppm to less than 5 ppm within 30 min. A significant decrease in the electrical conductivity, acidity, pH, TSS, and yellowness, but a significant increase in the lightness, of the juice was observed upon ED. Concentrations of almost all amino acids of the ED-treated juice significantly decreased. The concentrations of 8 major compound contributors to the pineapple aroma also significantly decreased. Adding the pulp back to the ED-treated juice increased the amino acids concentrations; however, it led to a significant decrease in the concentrations of the aroma compounds.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methyl 3-(methylthio)propionate, ≥98%, FG
Sigma-Aldrich
Methyl 3-(methylthio)propionate, 98%
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Ethyl trans-3-hexenoate, ≥98%, FG
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)