Merck
  • Variation of levels and distribution of N-nitrosamines in different seasons in drinking waters of East China.
Variation of levels and distribution of N-nitrosamines in different seasons in drinking waters of East China.
Environmental science and pollution research international (2015-04-11)
Ting Li, Dian Yu, Qiming Xian, Aimin Li, Cheng Sun
ABSTRACT
We surveyed the occurrence of nine N-nitrosamine species in ten bottled drinking waters from supermarket and other water samples including raw waters, finished waters, and distribution system waters from nine municipal drinking water treatment plants in eight cities of Jiangsu Province, East China. N-nitrosodimethylamine (NDMA) was detected in one of ten bottled drinking water samples at concentration of 4.8 ng/L and N-nitrosomorpholine (NMor) was detected in four of the ten bottles with an average concentration and a standard deviation of 16 ± 15 ng/L. The levels of nitrosamines in the distribution system water samples collected during summer season ranged from below detection limit (BDL) to 5.4 ng/L for NDMA, BDL to 9.5 ng/L for N-nitrosomethylethylamine (NMEA), BDL to 2.7 ng/L for N-nitrosodiethylamine (NDEA) and BDL to 8.5 ng/L for N-nitrosopyrrolidine (NPyr). Samples of distribution system waters collected in winter season had levels of nitrosamines ranged from BDL to 45 ng/L for NDMA, BDL to 5.2 ng/L for NPyr, and BDL to 309 ng/L for N-nitrosopiperidine (NPip). A positive correlation of the concentration of NDMA as well as the total nine N-nitrosamines between finished waters and distribution system waters was observed. Both dissolved organic carbon and nitrite were found to correlate linearly with N-nitrosamine levels in raw waters.
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Dichloromethane-d2, "100%", 99.96 atom % D
Sigma-Aldrich
Dichloromethane-d2, 99.9 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
N-Ethylmethylamine, 97%
Sigma-Aldrich
Dichloromethane-d2, ≥99.5 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dichloromethane-d2, 99.9 atom % D
Sigma-Aldrich
Dichloromethane-d2, 99.5 atom % D