Merck
  • The effect of transient, moderate dietary phosphorus deprivation on phosphorus metabolism, muscle content of different phosphorus-containing compounds, and muscle function in dairy cows.
The effect of transient, moderate dietary phosphorus deprivation on phosphorus metabolism, muscle content of different phosphorus-containing compounds, and muscle function in dairy cows.
Journal of dairy science (2015-06-01)
W Grünberg, P Scherpenisse, P Dobbelaar, M J Idink, I D Wijnberg
ABSTRACT
Hypophosphatemia is a common finding in periparturient and anorectic cattle. Although the clinical relevance of hypophosphatemia in cattle is uncertain, it has been empirically associated with persistent recumbency, specifically in periparturient dairy cows. The objective of the present study was to determine if transient dietary phosphorus (P) deprivation over a course of 5 wk, by feeding an approximately 40% P-deficient ration to lactating dairy cows, would result in altered muscle function or muscle P metabolism severe enough to present a risk for animal health and well-being. In addition, we wanted to determine the association between the plasma phosphate concentration ([Pi]) and muscle tissue P content to assess to what extent intracellular P deprivation of muscle cells could be extrapolated from subnormal plasma [Pi]. Ten healthy multiparous, mid-lactating dairy cows received a ration with a P content of 0.18% over a period of 5 wk. Following the P-deprivation phase, the same ration supplemented with P to obtain a dietary P content of 0.43% was fed for 2 wk. Blood and urine samples were collected regularly and muscle biopsies were obtained repeatedly to determine the P content in muscle tissue. Function of skeletal and heart muscles was evaluated by electrocardiography and electromyography conducted repeatedly throughout the study. Feeding the P-deficient ration resulted in the rapid development of marked hypophosphatemia. The lowest plasma [Pi] were measured after 9 d of P depletion and were, on average, 60% below predepletion values. Plasma [Pi] increased thereafter, despite ongoing dietary P depletion. None of the animals developed clinical signs commonly associated with hypophosphatemia or any other health issues. Urine analysis revealed increasing renal calcium, pyridinoline, and hydroxypyridinoline excretion with ongoing P deprivation. Biochemical muscle tissue analysis showed that dietary P depletion and hypophosphatemia were not associated with a decline in muscle tissue P content. Electromyographic examination revealed increased occurrence of pathological spontaneous activity in striated muscles after 2 wk of dietary P depletion in several cows, which could be suggestive of neuromuscular membrane instability. No effect on heart muscle activity was identified electrocardiographically. These results suggest that counter-regulatory mechanisms were sufficient to maintain normal muscle tissue P content during transient and moderate P deprivation. Muscle function was not grossly affected, although the increased occurrence of pathological spontaneous activity suggests that subclinical neuropathy or myopathy, or both, may have occurred with ongoing P deprivation. The results presented here indicate that plasma [Pi] is unsuitable for assessing muscle tissue P content in cattle.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications