Merck
  • Determining the location and nearest neighbours of aluminium in zeolites with atom probe tomography.
Determining the location and nearest neighbours of aluminium in zeolites with atom probe tomography.
Nature communications (2015-07-03)
Daniel E Perea, Ilke Arslan, Jia Liu, Zoran Ristanović, Libor Kovarik, Bruce W Arey, Johannes A Lercher, Simon R Bare, Bert M Weckhuysen
ABSTRACT
Zeolite catalysis is determined by a combination of pore architecture and Brønsted acidity. As Brønsted acid sites are formed by the substitution of AlO4 for SiO4 tetrahedra, it is of utmost importance to have information on the number as well as the location and neighbouring sites of framework aluminium. Unfortunately, such detailed information has not yet been obtained, mainly due to the lack of suitable characterization methods. Here we report, using the powerful atomic-scale analysis technique known as atom probe tomography, the quantitative spatial distribution of individual aluminium atoms, including their three-dimensional extent of segregation. Using a nearest-neighbour statistical analysis, we precisely determine the short-range distribution of aluminium over the different T-sites and determine the most probable Al-Al neighbouring distance within parent and steamed ZSM-5 crystals, as well as assess the long-range redistribution of aluminium upon zeolite steaming.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium nitrate, suitable for plant cell culture
Sigma-Aldrich
Ammonium nitrate, BioXtra, ≥99.5%