Merck
  • Effect of grinding intensity and pelleting of the diet on indoor particulate matter concentrations and growth performance of weanling pigs.
Effect of grinding intensity and pelleting of the diet on indoor particulate matter concentrations and growth performance of weanling pigs.
Journal of animal science (2014-12-31)
T Ulens, P Demeyer, B Ampe, H Van Langenhove, S Millet
ABSTRACT
This study evaluated the effect of feed form and grinding intensity of the pig diet and the interaction between both on the particulate matter (PM) concentrations inside a pig nursery and the growth performances of weanling pigs. Four diets were compared: finely ground meal, coarsely ground meal, finely ground pellets, and coarsely ground pellets. Four weaning rounds with 144 pigs per weaning round, divided over 4 identical compartments, were monitored. Within each weaning round, each compartment was randomly assigned to 1 of 4 treatments. A hammer mill with a screen of 1.5 or 6 mm was used to grind the ingredients of the finely ground and coarsely ground feeds, respectively. Indoor concentrations of the following PM fractions were measured: PM that passes through a size-selective inlet with a 50 % efficiency cutoff at 10 (PM10) , 2.5 (PM2.5), or 1 (PM1) μm aerodynamic diameter, respectively (USEPA, 2004). Feeding pelleted diets instead of meal diets gave rise to higher PM10 (P < 0.001), PM2.5 (P < 0.001), and PM1 (P < 0.001) concentrations. Grinding intensity had an effect only on PM10 (P < 0.05) concentrations. No interaction between feed form and grinding intensity was found for any of the PM fractions. Interactions (P < 0.05) between feed form and grinding intensity on ADFI and ADG were found. Grinding intensity had an effect only on the meal diets with higher ADFI for the coarsely ground meal. Pigs fed the finely ground meal had a lower (P < 0.001) ADG than the other 3 diets. Feed efficiency was influenced only by the feed form (P < 0.001) and not by the grinding intensity. Pelleting the feed gave rise to a higher G:F. In conclusion, a contradiction between environmental concerns and performance results was found. Feeding pelleted diets to the piglets improved growth performance but also increased indoor PM concentrations.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Citric acid monohydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, E330, 99.5-100.5% (based on anhydrous substance), grit
Sigma-Aldrich
Citric acid monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Citric acid monohydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O