Merck
  • In vitro metabolic conversion of the organic breakdown products of glucosinolate to goitrogenic thiocyanate anion.
In vitro metabolic conversion of the organic breakdown products of glucosinolate to goitrogenic thiocyanate anion.
Journal of the science of food and agriculture (2014-10-02)
Juyoung Lee, Hoonjeong Kwon
ABSTRACT
Glucosinolates are abundant in Brassicaceae vegetables, and they are degraded into various organic breakdown products (BPs) (R-CN, -NCS and -SCN) by myrosinase when plant tissues are damaged. This study was designed to investigate whether these BPs could be broken further into goitrogenic thiocyanate anions (SCN(-) ) metabolically and/or spontaneously. Ten glucosinolates were chosen for this study based on the various structures of their side chains. SCN(-) and cyanide anions (CN(-) ) liberated from the BPs of these glucosinolates were quantified after incubation with human liver S9 and rhodanese. Upon treatment with metabolic enzymes, CN(-) was produced from all organic thiocyanates, aliphatic and benzyl nitriles, then a substantial amount of produced CN(-) was further metabolized to SCN(-) by rhodanese. All organic thiocyanates and allyl isothiocyanate were metabolized to produce SCN(-), without involving CN(-) production. Spontaneous degradation to SCN(-) in an aqueous environment was observed only in 4-(methylthio)butyl thiocyanate, though the enzymatic reaction rate exceeded the spontaneous one. Among these BPs, the major source of SCN(-) was organic thiocyanates. The results show that some organic nitriles, organic thiocyanates and allyl isothiocyanate may be regarded as potential sources of SCN(-) through metabolism when people ingest glucosinolate-containing vegetables.
Sigma-Aldrich
L-Sulforaphane, ≥95% (HPLC), oil
Sigma-Aldrich
Ethyl isothiocyanate, 97%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Butyronitrile, ≥99%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Allyl isothiocyanate, ≥95%, FCC
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Benzyl isothiocyanate, 98%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
3-Phenylpropionitrile, 99%
Sigma-Aldrich
Butyl isothiocyanate, 99%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Benzyl thiocyanate, ≥95.0% (GC)
Sigma-Aldrich
Phenethyl isothiocyanate, 99%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Allyl isothiocyanate, 95%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
3-Indoleacetonitrile, 98%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Benzyl cyanide, 98%
Sigma-Aldrich
Propionitrile, 99%
Sigma-Aldrich
Allyl cyanide, 98%
Sigma-Aldrich
Valeronitrile, 99.5%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
2-Phenylethyl isothiocyanate, FG