Merck
  • Solid state compatibility study and characterization of a novel degradation product of tacrolimus in formulation.
Solid state compatibility study and characterization of a novel degradation product of tacrolimus in formulation.
Journal of pharmaceutical and biomedical analysis (2015-03-26)
Tanja Rozman Peterka, Rok Grahek, Jure Hren, Andrej Bastarda, Jure Bergles, Uroš Urleb
ABSTRACT
Tacrolimus is macrolide drug that is widely used as a potent immunosuppressant. In the present work compatibility testing was conducted on physical mixtures of tacrolimus with excipients and on compatibility mixtures prepared by the simulation of manufacturing process used for the final drug product preparation. Increase in one major degradation product was detected in the presence of magnesium stearate based upon UHPLC analysis. The degradation product was isolated by preparative HPLC and its structure was elucidated by NMR and MS studies. Mechanism of the formation of this degradation product is proposed based on complementary degradation studies in a solution and structural elucidation data. The structure was proven to be alpha-hydroxy acid which is formed from the parent tacrolimus molecule through a benzilic acid type rearrangement reaction in the presence of divalent metallic cations. Degradation is facilitated at higher pH values.
Sigma-Aldrich
Magnesium citrate, AldrichCPR
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Calcium hydroxide, 99.995% trace metals basis
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG