Merck
PTPRO-mediated autophagy prevents hepatosteatosis and tumorigenesis.
Oncotarget (2015-04-01)
Wenjie Zhang, Jiajie Hou, Xiaochen Wang, Runqiu Jiang, Yin Yin, Jie Ji, Lei Deng, Xingxu Huang, Ke Wang, Beicheng Sun
ABSTRACT
Autophagy plays a critical role in the progression of nonalcoholic steatohepatitis (NASH) and hepatocellular carcinoma (HCC). Protein tyrosine phosphatase receptor type O (PTPRO) was recently identified as a tumor suppressor, but little is known about its role in NASH. Here, we investigated the role of PTPRO-dependent autophagy in insulin resistance, lipid metabolism, and hepatocarcinogenesis. Wild-type (WT) and ptpro-/- mice were fed a high-fat diet (HFD) for another 16 weeks after diethylnitrosamine (DEN) injection to induce NASH. Ptpro-/- mice exhibited severe liver injury, insulin resistance, hepatosteatosis and autophagy deficiency compared with WT littermates. PTPRO deletion also promoted the induction of lipogenic target genes and decreases in β-oxidation-related genes. Increased activation of AKT and accumulation of cytoplasmic p53 was detected in ptpro-/- mice, which in combination repressed autophagy. Intriguingly, hyperinsulinemia involving AKT activation was also exacerbated in HFD-fed mice due to PTPRO deletion. Activation of AKT induced stabilization of the MDMX/MDM2 heterocomplex, thus promoting p53 accumulation in the cytoplasm. Inhibition of AKT restored autophagy and p53 accumulation in hepatocytes, indicating that AKT acts upstream of p53. Due to hyperinsulinemia and autophagy deficiency, a HFD could aggravate steatohepatitis in ptpro-/- mice. Importantly, the expression of PTPRO was much decreased in human steatohepatitis, which was associated with increased p62 accumulation. Together, these data indicate that PTPRO regulates insulin and lipid metabolism via the PI3K/Akt/MDM4/MDM2/P53 axis by affecting autophagy.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Gey′s Balanced Salt Solution, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
N-Nitrosodiethylamine, ≥99.0% (GC)
Sigma-Aldrich
N-Nitrosodiethylamine, liquid
Sigma-Aldrich
N-Nitrosodiethylamine, ISOPAC®
Sigma-Aldrich
Anti-PTPRO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution