Merck
  • Okibacterium endophyticum sp. nov., a novel endophytic actinobacterium isolated from roots of Salsola affinis C. A. Mey.
Okibacterium endophyticum sp. nov., a novel endophytic actinobacterium isolated from roots of Salsola affinis C. A. Mey.
Antonie van Leeuwenhoek (2015-01-09)
Hong-Fei Wang, Yong-Guang Zhang, Li Li, Wei-Hong Liu, Wael N Hozzein, Ji-Yue Chen, Jian-Wei Guo, Yuan-Ming Zhang, Wen-Jun Li
ABSTRACT
A white bacterial strain, designated EGI 650022(T), was isolated from the roots of Salsola affinis C. A. Mey, collected from Urumqi City, Xinjiang, north-western China. The strain was found to be aerobic, Gram-stain positive, oxidase-positive and catalase-positive. Cells were non-motile and irregular rods. Growth occurred at NaCl concentrations between 0 and 7 % (w/v), temperatures between 5 and 45 °C, and pH 6.0-9.0. Phylogenetic analysis based on 16S rRNA gene sequence indicated that strain EGI 650022(T) belongs to a clade with the genera Okibacterium and Plantibacter in the family Microbacteriaceae. The novel strain EGI 650022(T) showed highest levels of 16S rRNA gene sequence similarity with members of the genera Okibacterium and Plantibacter (97.2-98.0 %). The cell-wall peptidoglycan contained glutamate, homoserine, glycine, alanine and lysine. The predominant menaquinones (MKs) were MK-11, MK-12 and MK-12 (H4). The polar lipid pattern comprised phosphatidylglycerol, diphosphatidylglycerol, two unknown glycolipids and two unknown phospholipids. The major fatty acids were anteiso-C15:0 and anteiso-C17:0. The DNA G+C content was 66.0 mol%. The DNA-DNA relatedness values of strain EGI 650022(T) with Okibacterium fritillariae DSM 12584(T), Plantibacter flavus DSM 14012(T) and Plantibacter auratus DSM 19586(T) were 39.7, 19.7 and 22.0 %. Based on phylogenetic, phenotypic, chemotaxonomic and DNA-DNA hybridization data, strain EGI 650022(T) is considered to represent a novel species of the genus Okibacterium, for which the name Okibacterium endophyticum sp. nov. is proposed; the type strain is EGI 650022(T) (=JCM 30086(T) = KCTC 29492(T)).
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Galactitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Mucic acid, 97%
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
D-(−)-Quinic acid, 98%
Supelco
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis
Supelco
Phosphomolybdic acid hydrate, for microscopy
Sigma-Aldrich
D-Aspartic acid, ReagentPlus®, 99%
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Sigma-Aldrich
L-(−)-Fucose, ≥98% (GC)
Sigma-Aldrich
D-(+)-Fucose, ≥97% (GC)
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)
Sigma-Aldrich
Phosphomolybdic Acid Solution
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Phosphomolybdic acid solution, ready-to-use spray and plunge reagent, for chromatography
USP
Galactitol, United States Pharmacopeia (USP) Reference Standard
Supelco
N-Acetylglucosamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphomolybdic acid hydrate, ACS reagent
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
meso-Erythritol, ≥99% (GC)