Merck
  • Expressional changes of genes and miRNA in common megakaryocyte-erythroid progenitors from lower-risk myelodysplastic syndrome.
Expressional changes of genes and miRNA in common megakaryocyte-erythroid progenitors from lower-risk myelodysplastic syndrome.
International journal of hematology (2014-07-25)
Kazuhiro Maki, Ko Sasaki, Yasunobu Nagata, Fusako Nagasawa, Yuka Nakamura, Seishi Ogawa, Kinuko Mitani
ABSTRACT
Myelodysplastic syndrome (MDS) is a stem cell tumor characterized by dysplastic features and ineffective hematopoiesis in the early phase and leukemic progression in the late phase. Speculating that differences in the expression of genes and microRNA (miRNA) in control and MDS-derived erythroid progenitors may cause ineffective erythropoiesis, we sorted common megakaryocyte-erythroid progenitors (MEPs) in bone marrow cells from three lower-risk MDS patients, and compared expression levels of genes and miRNA with those from controls. In apoptosis-related pathways, the expression of some pro-apoptotic genes, such as cell death-inducing DFFA-like effector A, caspase 5, and Fas ligand, was elevated in MDS-derived MEPs, while those of anti-apoptotic CD40 and tumor necrosis factor were lower. In hematopoiesis-regulating pathways, RUNX1 and ETV6 genes showed reduced expression. Expression profiling revealed that three and 35 miRNAs were significantly up- and down-regulated in MDS-derived MEPs. MIR9 exhibited robust expression in MEPs and CD71+GlyA+ erythroid cells derived from one of the three patients. Interestingly, overexpression of MIR9 inhibited the accumulation of hemoglobin in UT-7/GM cells. Some of these alterations in gene and miRNA expression may contribute to the pathogenesis of ineffective hematopoiesis in lower-risk MDS and provide molecular markers for sub-classification and making a prognosis.
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
EPO human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor human, GM-CSF, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Macrophage Colony-Stimulating Factor human, M-CSF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture