Merck
  • Activated innate lymphoid cells are associated with a reduced susceptibility to graft-versus-host disease.
Activated innate lymphoid cells are associated with a reduced susceptibility to graft-versus-host disease.
Blood (2014-05-24)
J Marius Munneke, Andreas T Björklund, Jenny M Mjösberg, Karin Garming-Legert, Jochem H Bernink, Bianca Blom, Cynthia Huisman, Marinus H J van Oers, Hergen Spits, Karl-Johan Malmberg, Mette D Hazenberg
ABSTRACT
Allogeneic hematopoietic stem cell transplantation (HSCT) is widely used to treat hematopoietic cell disorders but is often complicated by graft-versus-host disease (GVHD), which causes severe epithelial damage. Here we have investigated longitudinally the effects of induction chemotherapy, conditioning radiochemotherapy, and allogeneic HSCT on composition, phenotype, and recovery of circulating innate lymphoid cells (ILCs) in 51 acute leukemia patients. We found that reconstitution of ILC1, ILC2, and NCR(-)ILC3 was slow compared with that of neutrophils and monocytes. NCR(+) ILC3 cells, which are not present in the circulation of healthy persons, appeared both after induction chemotherapy and after allogeneic HSCT. Circulating patient ILCs before transplantation, as well as donor ILCs after transplantation, expressed activation (CD69), proliferation (Ki-67), and tissue homing markers for gut (α4β7, CCR6) and skin (CCR10 and CLA). The proportion of ILCs expressing these markers was associated with a decreased susceptibility to therapy-induced mucositis and acute GVHD. Taken together, these data suggest that ILC recovery and treatment-related tissue damage are interrelated and affect the development of GVHD.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Anti-IL7R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-KIT (Ab-721) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)