Merck
  • Evaluation of a rapid method for the therapeutic drug monitoring of aliskiren, enalapril and its active metabolite in plasma and urine by UHPLC-MS/MS.
Evaluation of a rapid method for the therapeutic drug monitoring of aliskiren, enalapril and its active metabolite in plasma and urine by UHPLC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-16)
Sylwia Magiera, Jacek Kusa
ABSTRACT
Given the increasing popularity of aliskiren, particularly in combination with angiotensin converting enzyme inhibitor (e.g. enalapril), it is important to determine whether its use in combination with these agents is associated with potentially life threatening safety events. Analytical methods for the simultaneous determination of both drugs in plasma and urine utilized in clinical studies on efficacy and safety have not been fully described in the literature. In this work, a new, fast and reliable method using a digitally controlled microextraction by packed sorbent (eVol(®)-MEPS) followed by ultra-high performance liquid chromatography (UHPLC) coupled with tandem mass spectrometry (MS/MS) was developed and validated to quantify an aliskiren, enalapril and its active metabolite in both human plasma and urine. Chromatographic separation was accomplished on a Poroshell 120 EC-C18 column with a gradient elution system consisting of 0.1% formic acid in water and acetonitrile (1.5min of total analysis). Detection was performed by multiple reaction monitoring (MRM) mode using electrospray ionization in the positive ion mode. This assay method has been fully validated in terms of selectivity, linearity, accuracy, precision, stability, recovery and matrix effect. The developed method can be applied to the routine determination of selected compounds in human plasma and urine and can be useful to elucidate the mechanisms of the potential risks triggered by the combination of aliskiren and enalapril as well as its active metabolite enalaprilat.
Sigma-Aldrich
Enalaprilat dihydrate, ≥98% (HPLC)
Supelco
Enalaprilat, analytical standard
USP
Enalaprilat, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Enalaprilat dihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)