Merck
  • Novel drug candidates for the treatment of metastatic colorectal cancer through global inverse gene-expression profiling.
Novel drug candidates for the treatment of metastatic colorectal cancer through global inverse gene-expression profiling.
Cancer research (2014-07-20)
Vera van Noort, Sebastian Schölch, Murat Iskar, Georg Zeller, Kristina Ostertag, Christine Schweitzer, Kristin Werner, Jürgen Weitz, Moritz Koch, Peer Bork
ABSTRACT
Drug-induced gene-expression profiles that invert disease profiles have recently been illustrated to be a starting point for drug repositioning. In this study, we validate this approach and focus on prediction of novel drugs for colorectal cancer, for which there is a pressing need to find novel antimetastatic compounds. We computationally predicted three novel and still unknown compounds against colorectal cancer: citalopram (an antidepressant), troglitazone (an antidiabetic), and enilconazole (a fungicide). We verified the compounds by in vitro assays of clonogenic survival, proliferation, and migration and in a subcutaneous mouse model. We found evidence that the mode of action of these compounds may be through inhibition of TGFβ signaling. Furthermore, one compound, citalopram, reduced tumor size as well as the number of circulating tumor cells and metastases in an orthotopic mouse model of colorectal cancer. This study proposes citalopram as a potential therapeutic option for patients with colorectal cancer, illustrating the potential of systems pharmacology.
Product Description
Supelco
Imazalil, PESTANAL®, analytical standard
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Enilconazole, European Pharmacopoeia (EP) Reference Standard