Merck
  • Resveratrol Suppresses Expression of VEGF by Human Retinal Pigment Epithelial Cells: Potential Nutraceutical for Age-related Macular Degeneration.
Resveratrol Suppresses Expression of VEGF by Human Retinal Pigment Epithelial Cells: Potential Nutraceutical for Age-related Macular Degeneration.
Aging and disease (2014-04-15)
Chandrasekharam N Nagineni, Raghavan Raju, Krishnasai K Nagineni, Vijay K Kommineni, Aswini Cherukuri, R Krishnan Kutty, John J Hooks, Barbara Detrick
ABSTRACT
Age-related macular degeneration (AMD) is a sight threating retinal eye disease that affects millions of aging individuals world-wide. Choroid-retinal pigment epithelium (RPE)-neuroretina axis in the posterior compartment of the eye is the primary site of AMD pathology. There are compelling evidence to indicate association of vascular endothelial growth factors (VEGF) to AMD. Here, we report the inhibitory actions of resveratrol (RSV) on inflammatory cytokine, TGF-β and hypoxia induced VEGF secretion by human retinal pigment epithelial cells (HRPE). HRPE cultures prepared from aged human donor eyes were used for the studies in this report. HRPE secreted both VEGF-A and VEGF-C in small quantities constitutively. Stimulation with a mixture of inflammatory cytokines (IFN-γ, TNF-α, IL-1β), significantly increased the secretion of both VEGF-A and VEGF-C. RSV, in a dose dependent (10-50 uM) manner, suppressed VEGF-A and VEGF-C secretion induced by inflammatory cytokines significantly. RT-PCR analysis indicated that effects of RSV on VEGF secretion were possibly due to decreased mRNA levels. TGF-β and cobalt chloride (hypoxia mimic) also upregulated HRPE cell production of VEGF-A, and this was inhibited by RSV. In contrast, RSV had no effect on anti-angiogenic molecules, endostatin and pigment epithelial derived factor secretion. Studies using an in vitro scratch assay revealed that wound closure was also inhibited by RSV. These results demonstrate that RSV can suppress VEGF secretion induced by inflammatory cytokines, TGF-β and hypoxia. Under pathological conditions, over expression of VEGF is known to worsen AMD. Therefore, RSV may be useful as nutraceutical in controlling pathological choroidal neovascularization processes in AMD.
Sigma-Aldrich
Human VEGF-C ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Resveratrol, analytical standard
Resveratrol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland