Merck
  • Cisplatin-induced duplex dissociation of complementary and destabilized short GG-containing duplex RNAs.
Cisplatin-induced duplex dissociation of complementary and destabilized short GG-containing duplex RNAs.
Dalton transactions (Cambridge, England : 2003) (2014-06-27)
Christopher Polonyi, Alak Alshiekh, Lamya A Sarsam, Maria Clausén, Sofi K C Elmroth
ABSTRACT
The ability of the anticancer active drug cisplatin to exert biological activity through interference with nucleic acid function is well documented. Since kinetics play a key role in determining product distributions in these systems, methods for accurate documentation of reactivity serve the purpose to identify preferential metal binding sites. In the present study, the aim has been to further explore a recently communicated approach (C. Polonyi and S. K. C. Elmroth, J. Chem. Soc., Dalton Trans., 2013, 42, 14959-14962) utilizing UV/vis spectroscopy and metal induced duplex RNA melting for monitoring of kinetics. More specifically, the sensitivity of the UV/vis-methodology has been evaluated by investigation of how overall length and changes of base-pairing in the close vicinity of a centrally located GG-site affect the rate of cisplatin binding, using the intracellularly active mono-aquated form of cisplatin (cis-Pt(NH3)2Cl(OH2)(+), ()) as the platination reagent. For this purpose, the reactivity of five different 13- to 17 base-pair duplex RNAs was monitored at 38 °C. A common trend of a ca. 10-fold reduction in reactivity was found to accompany an increase of bulk sodium concentration from CNa+ = 122 mM to 1.0 M. Typical half-lives are exemplified by the interaction of with the fully complementary 15-mer RNA-1 with t1/2 = ca. 0.5 and 4.8 hours, at CNa+ = 122 mM and 1.0 M respectively, and C = 45 μM. Lowering of melting temperature (Tm) was found to promote reactivity regardless of whether the change involved a decrease or increase of the RNA length. For example, at CNa+ = 1.0 M, truncation of the fully complementary and GG-containing 15-mer RNA-1 (Tm = 68.9 °C) to the 13-mer RNA-1-1-S (Tm = 63.9 °C) resulted in an increase of k2,app from ca. 0.9 M(-1) s(-1) to 2.0 M(-1) s(-1). Further, the 17-mer RNA-1-4 (Tm = 42.0 °C) with a central U4 bulge exhibited the highest reactivity of the sequences studied with k2,app = 4.0 M(-1) s(-1). The study shows that the reactivity of GG-sequences in RNA exhibit a strong variation depending on exact sequence context, and with imperfectly matched and/or stacked regions as particularly reactive sites.
Product Description
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium perchlorate hydrate, 99.99% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioXtra, ≥98.0% (T)
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, puriss. p.a., ACS reagent, ≥99.0% (T)
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, puriss. p.a., reag. Ph. Eur., 98.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, meets analytical specification of Ph. Eur., BP, 98.5-101% (calc. to the dried substance)
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium perchlorate monohydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Sodium perchlorate monohydrate, ACS reagent