Merck
  • In-syringe dispersive solid-phase extraction using dissolvable layered double oxide hollow spheres as sorbent followed by high-performance liquid chromatography for determination of 11 phenols in river water.
In-syringe dispersive solid-phase extraction using dissolvable layered double oxide hollow spheres as sorbent followed by high-performance liquid chromatography for determination of 11 phenols in river water.
Journal of chromatography. A (2014-12-02)
Sheng Tang, Xuan Hao Lin, Sam Fong Yau Li, Hian Kee Lee
ABSTRACT
Layered double oxide hollow spheres (LDO-HSs) were synthesized and employed as a dissolvable sorbent in dispersive solid-phase extraction (DSPE) to extract eleven United States Environmental Protection Agency's priority phenols from aqueous samples. With their higher specific surface area, LDO-HSs showed much higher extraction efficiency than normal layered double hydroxides and layered double oxides. The entire extraction process was accomplished in a syringe. After DSPE, the sorbent with the analytes was isolated conveniently by directly expelling the spent sample solution out of the syringe. The analyte-enriched sorbent was then subsequently dissolved by withdrawing an acidic solution into the syringe. The final extract was analyzed by high-performance liquid chromatography with ultraviolet detection. The results showed that this method provided low limits of detection for the phenols (0.005-0.153 μg/L), good linearity (r(2)≥ 0.9956) and relative standard deviations of ≤ 6.7%. The optimized method was applied to water samples from 3 rivers. This simple extraction procedure was demonstrated to be a fast, efficient and convenient DSPE approach.
Sigma-Aldrich
Magnesium nitrate hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Magnesium nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Magnesium nitrate hexahydrate, BioXtra, ≥98%
Sigma-Aldrich
Magnesium nitrate hexahydrate, puriss. p.a., ACS reagent, 98.0-102.0% (KT)
Sigma-Aldrich
Magnesium nitrate hexahydrate, ACS reagent, 99%
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%