Merck
  • Site specific solubility improvement using solid dispersions of HPMC-AS/HPC SSL--mixtures.
Site specific solubility improvement using solid dispersions of HPMC-AS/HPC SSL--mixtures.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-04-29)
Damir Elmar Zecevic, Robin Meier, Rolf Daniels, Karl-Gerhard Wagner
ABSTRACT
Many upcoming drug candidates are pH-dependent poorly soluble weak bases in the pH range of the gastrointestinal tract. This often leads to a high in vivo variability and bioavailability issues. Aiming to overcome these limitations, the design of solid dispersions for site specific dissolution improvement or maintenance of a potent supersaturation over the entire gastro-intestinal pH-range, is proposed to assure a reliable drug therapy. Solid dispersions containing different ratios of Dipyridamole (DPD) or Griseofulvin (GRI) and the enteric polymer hydroxypropylmethylcellulose-acetate succinate (HPMC-AS) and the water soluble low-viscosity hydroxypropylcellulose (HPC-SSL) were prepared by hot melt extrusion (HME). The solid dispersions were evaluated for their solid state, dissolution characteristics applying a three pH-step dissolution method following an acidic to neutral pH transition and stability. The use of HPMC-AS in binary mixtures with DPD and GRI facilitated increased solubility and supersaturation at pH-controlled release of the preserved amorphous state of the dispersed drug, which even inverted the pH-dependent solubility profile of the weakly basic model drug (Dipyridamole). I.e. a potent site specific delivery system was created. With ternary solid dispersions of API, HPMC-AS and HPC-SSL, tailored release profiles with superior supersaturation over the applied pH-range could be obtained. At the same time, binary and ternary mixtures showed favorable stability properties at a temperature difference between glass transition temperature and the applied storage temperature of down to 16°C.
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%
Sigma-Aldrich
Dipyridamole, ≥98% (HPLC)
Dipyridamole, European Pharmacopoeia (EP) Reference Standard
USP
Griseofulvin, United States Pharmacopeia (USP) Reference Standard
Supelco
Griseofulvin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Griseofulvin Permeability Diameter, United States Pharmacopeia (USP) Reference Standard
Dipyridamole for peak identification, European Pharmacopoeia (EP) Reference Standard