Merck
  • Fingerprinting of vegetable oil minor components by multidimensional comprehensive gas chromatography with dual detection.
Fingerprinting of vegetable oil minor components by multidimensional comprehensive gas chromatography with dual detection.
Analytical and bioanalytical chemistry (2014-09-12)
Giorgia Purcaro, Laura Barp, Marco Beccaria, Lanfranco S Conte
ABSTRACT
The potentiality of a multidimensional comprehensive gas chromatographic (GC × GC) method, employing a simultaneous dual detection (FID and mass spectrometer), to generate peculiar two-dimensional chromatograms to be used as a chemical fingerprint, was investigated to characterize minor compounds in edible oil, particularly olive oil. The best column combination for this application was investigated comparing two column sets (orthogonal or reverse-type), equivalent in terms of theoretical plate number, but differing in stationary phase combination. The apolar × mid-polar set gave a superior separation power, thus was used for further characterization. Different levels of information were extrapolated from the two-dimensional chromatogram. Using the FID, reliable quantification of the alkyl esters fatty acids and waxes was obtained, comparable to the results obtained using the official method, as required by the European legislation. However, thanks to a slight modification of the sample preparation method, the increased separation power obtained using the GC × GC method, and the support of the mass spectrometer detector, further diagnostic information was extrapolated considering the free sterol and tocopherol fractions. In particular, the profiles of extra virgin olive oil samples were compared with a hazelnut oil sample, highlighting that the latter was characterized by a larger number of compounds, completely absent in the extra virgin olive oil samples, which can be used to detect illegal admixtures.
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Heptane, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Supelco
Heptane, analytical standard
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Dodecyl arachidate, ≥99%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
Metolachlor OA, PESTANAL®, analytical standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide, for GC derivatization, LiChropur, ≥99.0%