Merck
  • Non-destructive micro-analytical differentiation of copper pigments in paint layers of works of art using laboratory-based techniques.
Non-destructive micro-analytical differentiation of copper pigments in paint layers of works of art using laboratory-based techniques.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2014-06-04)
Silvie Svarcová, Zdeňka Cermáková, Janka Hradilová, Petr Bezdička, David Hradil
ABSTRACT
An unambiguous identification of pigments in paint layers of works of art forms a substantial part of the description of a painting technique, which is essential for the evaluation of the work of art including determination of the period and/or region of its creation as well as its attribution to a workshop or an author. Copper pigments represent a significant group of materials used in historic paintings. Because of their substantial diversity and, on the other hand, similarity, their identification and differentiation is a challenging task. An analytical procedure for unambiguous determination of both mineral-type (azurite, malachite, posnjakite, atacamite, etc.) and verdigris-type (copper acetates) copper pigments in the paint layers is presented, including light microscopy under VIS and UV light, electron microscopy with elemental microanalysis, Fourier transformed infrared micro-spectroscopy (micro-FTIR), and X-ray powder micro-diffraction (micro-XRD). Micro-Raman measurements were largely hindered by fluorescence. The choice of the analytical methods meets the contemporary requirement of a detailed description of various components in heterogeneous and minute samples of paint layers without their destruction. It is beneficial to use the combination of phase sensitive methods such as micro-FTIR and micro-XRD, because it allows the identification of both mineral-type and verdigris-type copper pigments in one paint layer. In addition, preliminary results concerning the study of the loss of crystallinity of verdigris-type pigments in proteinaceous binding media and the effect of lead white and lead tin yellow as highly absorbing matrix on verdigris identification in paint layers are reported.
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Germanium, microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Germanium, rod, 6mm, diameter 6.0mm, single crystal, 100%
Germanium, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, sheet, 50x50mm, thickness 3.0mm, polycrystalline, 99.999%
Germanium, disks, 20mm, thickness 1.0mm, single crystal, 100%
Germanium, sheet, 10x10mm, thickness 0.25mm, polycrystalline, 99.999%
Germanium, sheet, 6x6mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, microfoil, 25x25mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Germanium, sheet, 10x10mm, thickness 0.5mm, single crystal, -111, 100%
Germanium, sheet, 7x24mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, microfoil, 25x25mm, thinness 0.5μm, specific density 333μg/cm2, 6 micron aluminum permanent support, 100%
Germanium, sheet, 10x10mm, thickness 0.6mm, single crystal, -111, 100%
Germanium, rod, 25mm, diameter 2.0mm, polycrystalline, n-type, 99.999%
Germanium, sheet, 25x25mm, thickness 1.0mm, single crystal, 99.999%
Germanium, sheet, 50x50mm, thickness 0.5mm, single crystal, 99.999%
Germanium, sheet, 25x25mm, thickness 3.0mm, polycrystalline, 99.999%
Germanium, rod, 50mm, diameter 5mm, polycrystalline, n-type, 99.999%
Germanium, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
Germanium, disks, 15mm, thickness 1.0mm, polycrystalline, 100%