Merck
  • An enhanced LSPR fiber-optic nanoprobe for ultrasensitive detection of protein biomarkers.
An enhanced LSPR fiber-optic nanoprobe for ultrasensitive detection of protein biomarkers.
Biosensors & bioelectronics (2014-05-27)
Mollye Sanders, Yongbin Lin, Jianjun Wei, Taylor Bono, Robert G Lindquist
ABSTRACT
A miniaturized, localized surface plasmon resonance (LSPR)-coupled fiber-optic (FO) nanoprobe is reported as a biosensor that is capable of label-free, sensitive detection of a cancer protein biomarker, free prostate specific antigen (f-PSA). The biosensor is based on the LSPR at the reusable dielectric-metallic hybrid interface with a robust, gold nano-disk array at the fiber end facet that is directly fabricated using EBL and metal lift-off process. The f-PSA has been detected with a mouse anti-human PSA monoclonal antibody (mAb) as a specific receptor linked with a self-assembled monolayer at the LSPR-FO facet surfaces. Experimental investigation and data analysis found near field refractive index (RI) sensitivity at ~226 nm/RIU with current LSPR-FO nanoprobe, and demonstrated the lowest limit of detection (LOD) at 100 fg/mL (~3 fM) of f-PSA in PBS solutions. The control experimentation using 5mg/mL bovine serum albumin in PBS and nonspecific surface test shows the excellent specificity and selectivity in the detection of f-PSA in PBS. These results present important progress towards a miniaturized, multifunctional fiber-optic technology that integrates informational communication and sensing function for developing a high performance, label-free, point-of-care (POC) device.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
8-Mercapto-1-octanol, 98%
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)