Merck
  • Pharmacological evaluation of the anxiolytic-like effects of EMD 386088, a partial 5-HT6 receptor agonist, in the rat elevated plus-maze and Vogel conflict tests.
Pharmacological evaluation of the anxiolytic-like effects of EMD 386088, a partial 5-HT6 receptor agonist, in the rat elevated plus-maze and Vogel conflict tests.
Neuropharmacology (2014-06-07)
Magdalena Jastrzębska-Więsek, Agata Siwek, Anna Partyka, Monika Kubacka, Szczepan Mogilski, Anna Wasik, Marcin Kołaczkowski, Anna Wesołowska
ABSTRACT
The 5-HT6 is one of the most recent additions to the 5-HT receptor family. Its pharmacological profile and anatomical distribution is suggestive of a putative role in mood disorders. Most of preclinical evidence suggests an anxiolytic-like action of 5-HT6 receptor antagonists. Evaluation the anxiolytic-like effects of EMD 386088, a partial 5-HT6receptor agonist, and its putative mechanism of action in rats. EMD 386088, administered intraperitoneally at a dose of 2.5 mg/kg evoked specific anxiolytic-like activity in the automated version of the conflict drinking Vogel and the elevated plus-maze tests visible by increasing all parameters indicating a potential anti-anxiety effect. Its activity was blocked by the selective 5-HT6 receptor antagonist SB 271046, but not by the selective GABAA/benzodiazepine receptor antagonist flumazenil. EMD 386088 did not intensify an anxiolytic-like effect produced by diazepam in the elevated plus-maze test. These findings suggest that EMD 386088, a 5-HT6 receptor agonist, produces anxiolytic-like activity after systemic administration which may result from direct stimulation of 5-HT6 receptors.
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Flumazenil, >99% (HPLC), solid
Ondansetron hydrochloride dihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ondansetron hydrochloride dihydrate, ≥98% (HPLC), powder
Supelco
Ondansetron Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Ondansetron for TLC system suitability, European Pharmacopoeia (EP) Reference Standard
Ondansetron for LC system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Ondansetron hydrochloride, United States Pharmacopeia (USP) Reference Standard