Merck
  • N-arylated-lactam-type iminosugars as new immunosuppressive agents: discovery, optimization, and biological evaluation.
N-arylated-lactam-type iminosugars as new immunosuppressive agents: discovery, optimization, and biological evaluation.
Chemistry, an Asian journal (2014-04-05)
Xiaowei Wu, Fu-Yu Zhang, Jingjing Zhu, Chengcheng Song, De-Cai Xiong, Yifa Zhou, Yuxin Cui, Xin-Shan Ye
ABSTRACT
We have previously described the discovery of N-alkylated iminosugars that showed immunosuppressive activity both in vitro and in vivo. Herein, we report the synthesis and biological evaluation of N-arylated lactam-type iminosugar derivatives. The synthesis started from simple monosaccharides and featured a Buchwald-Hartwig coupling reaction to construct the key N-aryl connection, thereby providing a highly diverse compound library. Structure-activity relationship studies, guided by a mouse-spleen-proliferation assay, led to the identification of 'hit' compound 12 f. Subsequently, the systematic modification of compound 12 f afforded compounds 21 h, 21 k, 21 n, 21 t, and 21 x with improved activities (IC50 =12-30 μM) and low Jurkat cytotoxicities (IC50 >100 μM). These new compounds also inhibited the secretion of IFN-γ and IL-4, which are hallmark cytokines of Th1 and Th2 cells, respectively. This work demonstrated that the N-arylated iminosugar structure represents a new scaffold with immunosuppressive activity.
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Cotton Rat Ifng / Interferon Gamma ELISA Kit
Sigma-Aldrich
Mouse IFN-γ ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IFN-γ ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IFN-γ ELISA Kit, for serum, plasma, cell culture supernatant