Merck
  • Mononuclear cell secretome protects from experimental autoimmune myocarditis.
Mononuclear cell secretome protects from experimental autoimmune myocarditis.
European heart journal (2013-01-17)
Konrad Hoetzenecker, Matthias Zimmermann, Wolfram Hoetzenecker, Thomas Schweiger, Dagmar Kollmann, Michael Mildner, Balazs Hegedus, Andreas Mitterbauer, Stefan Hacker, Peter Birner, Christian Gabriel, Mariann Gyöngyösi, Przemyslaw Blyszczuk, Urs Eriksson, Hendrik Jan Ankersmit
ABSTRACT
Supernatants of serum-free cultured mononuclear cells (MNC) contain a mix of immunomodulating factors (secretome), which have been shown to attenuate detrimental inflammatory responses following myocardial ischaemia. Inflammatory dilated cardiomyopathy (iDCM) is a common cause of heart failure in young patients. Experimental autoimmune myocarditis (EAM) is a CD4+ T cell-dependent model, which mirrors important pathogenic aspects of iDCM. The aim of this study was to determine the influence of MNC secretome on myocardial inflammation in the EAM model. BALB/c mice were immunized twice with an alpha myosin heavy chain peptide together with Complete Freund adjuvant. Supernatants from mouse mononuclear cells were collected, dialysed, and injected i.p. at Day 0, Day 7, or Day 14, respectively. Myocarditis severity, T cell responses, and autoantibody formation were assessed at Day 21. The impact of MNC secretome on CD4+ T cell function and viability was evaluated using in vitro proliferation and cell viability assays. A single high-dose application of MNC secretome, injected at Day 14 after the first immunization, effectively attenuated myocardial inflammation. Mechanistically, MNC secretome induced caspase-8-dependent apoptosis in autoreactive CD4+ T cells. MNC secretome abrogated myocardial inflammation in a CD4+ T cell-dependent animal model of autoimmune myocarditis. This anti-inflammatory effect of MNC secretome suggests a novel and simple potential treatment concept for inflammatory heart diseases.
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, 99%
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%