Merck
  • The Cre/loxP system in Giardia lamblia: genetic manipulations in a binucleate tetraploid protozoan.
The Cre/loxP system in Giardia lamblia: genetic manipulations in a binucleate tetraploid protozoan.
International journal for parasitology (2014-04-22)
Petra B Wampfler, Carmen Faso, Adrian B Hehl
ABSTRACT
The bacteriophage-derived Cre/loxP system is a valuable tool that has revolutionised genetic and cell biological research in many organisms. We implemented this system in the intestinal parasite Giardia lamblia, an evolutionarily diverged protozoan whose binucleate and tetraploid genome organisation severely limits the application of reverse genetic approaches. We show that Cre-recombinase is functionally expressed in G. lamblia and demonstrate "recycling" of selectable markers. Providing the means for more complex and versatile genetic modifications, this technique massively increases the scope of functional investigations in G. lamblia and other protozoa with similar limitations with respect to genetic manipulation.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Pricing and availability is not currently available.
TPP® tissue culture plates, 24 well plate, flat bottom, polystyrene, 1.86 cm2, sterile, 126/cs
Pricing and availability is not currently available.