Merck
  • A randomized controlled trial of angiotensin-converting enzyme inhibition for skeletal muscle dysfunction in COPD.
A randomized controlled trial of angiotensin-converting enzyme inhibition for skeletal muscle dysfunction in COPD.
Chest (2014-02-22)
Dinesh Shrikrishna, Rebecca J Tanner, Jen Y Lee, Amanda Natanek, Amy Lewis, Patrick B Murphy, Nicholas Hart, John Moxham, Hugh E Montgomery, Paul R Kemp, Michael I Polkey, Nicholas S Hopkinson
ABSTRACT
Skeletal muscle impairment is a recognized complication of COPD, predicting mortality in severe disease. Increasing evidence implicates the renin-angiotensin system in control of muscle phenotype. We hypothesized that angiotensin-converting enzyme (ACE) inhibition would improve quadriceps function and exercise performance in COPD. This double-blind, randomized placebo-controlled trial investigated the effect of the ACE inhibitor, fosinopril, on quadriceps function in patients with COPD with quadriceps weakness. Primary outcomes were change in quadriceps endurance and atrophy signaling at 3 months. Quadriceps maximum voluntary contraction (QMVC), mid-thigh CT scan of the cross-sectional area (MTCSA), and incremental shuttle walk distance (ISWD) were secondary outcomes. Eighty patients were enrolled (mean [SD], 65 [8] years, FEV1 43% [21%] predicted, 53% men). Sixty-seven patients (31 fosinopril, 36 placebo) completed the trial. The treatment group demonstrated a significant reduction in systolic BP (Δ-10.5 mm Hg; 95% CI, -19.9 to -1.1; P = .03) and serum ACE activity (Δ-20.4 IU/L; 95% CI, -31.0 to -9.8; P < .001) compared with placebo. No significant between-group differences were observed in the primary end points of quadriceps endurance half-time (Δ0.5 s; 95% CI, -13.3-14.3; P = .94) or atrogin-1 messenger RNA expression (Δ-0.03 arbitrary units; 95% CI, -0.32-0.26; P = .84). QMVC improved in both groups (fosinopril: Δ1.1 kg; 95% CI, 0.03-2.2; P = .045 vs placebo: Δ3.6 kg; 95% CI, 2.1-5.0; P < .0001) with a greater increase in the placebo arm (between-group, P = .009). No change was shown in the MTCSA (P = .09) or ISWD (P = .51). This randomized controlled trial found that ACE inhibition, using fosinopril for 3 months, did not improve quadriceps function or exercise performance in patients with COPD with quadriceps weakness. Current Controlled Trials; No.: ISRCTN05581879; URL: www.controlled-trials.com.
Sigma-Aldrich
Fosinopril sodium, ≥98% (HPLC), powder