Merck
Inhibition of cytochrome P450 by ethambutol in human liver microsomes.
Toxicology letters (2014-06-10)
Sang Yoon Lee, Himchan Jang, Ji-Yoon Lee, Kwang-il Kwon, Soo Jin Oh, Sang Kyum Kim
ABSTRACT
Although cytochrome P450 inhibition is the major drug-drug interaction (DDI) mechanism in clinical pharmacotherapy, DDI of a number of well-established drugs have not been investigated. Rifampicin, isoniazid, pyrazinamide and ethambutol combination therapy inhibits clearance of theophylline in patients with tuberculosis. We determined the inhibitory effects of ethambutol on the activities of nine CYP isoforms including CYP1A2, 2A6, 2B6, 2C8, 2C9, 2C19, 2D6, 2E1 and 3A4 in pooled human liver microsomes (HLM). As measured by liquid chromatography-electrospray ionization tandem mass spectrometry, ethambutol exhibited strong inhibitory potential against CYP1A2 and CYP2E1, moderate against CYP2C19 and CYP2D6 and weak against CYP2A6, CYP2C9 and CYP3A4, based on the IC50 values. The K(i) value of ethambutol for CYP1A2 was 1.4 μM and for CYP2E1 was 2.9 μM. Inhibition of CYP1A2 and CYP2E1 was not increased by preincubation with ethambutol and β-nicotinamideadenine dinucleotide phosphate (NADPH), suggesting that the ethambutol-induced CYP inhibition may not be metabolism-dependent. Kinetic analysis showed that the inhibition of CYP1A2 and CYP2E1 by ethambutol was best fit to a competitive inhibition model. Formation of 1-methylxanthene and 1,3-dimethyluric acid from theophylline in HLM was decreased to 47% and 36%, respectively, by 3.0 μM ethambutol, which is comparable to its IC50 value against CYP1A2. Considering its maximal plasma concentrations of ~10 μM and long half-life of ~22 h, our findings raise the possibility that ethambutol causes significant DDIs in clinical situations with drugs with narrow therapeutic index, such as theophylline, in clinical situations.
Product Description
USP
Dextromethorphan, United States Pharmacopeia (USP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Ethambutol for system suitability, European Pharmacopoeia (EP) Reference Standard
Tolbutamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Furafylline, ≥98% (HPLC)
Supelco
Tolbutamide, analytical standard
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Supelco
Umbelliferone, analytical standard
Sigma-Aldrich
Chlorzoxazone
Sigma-Aldrich
Umbelliferone, 99%
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
USP
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Carbamazepine, powder