Merck
  • Saccharin mechanistic data and risk assessment: urine composition, enhanced cell proliferation, and tumor promotion.
Saccharin mechanistic data and risk assessment: urine composition, enhanced cell proliferation, and tumor promotion.
Pharmacology & therapeutics (1996-01-01)
J Whysner, G M Williams
ABSTRACT
Sodium saccharin (NaSac) produces bladder tumors consistently in male rats only after lifetime exposure that begins at birth. NaSac is not metabolized and is negative in most genotoxicity tests. NaSac-induced cell damage and proliferation have been proposed as important factors in tumor promotion, and dose-response information demonstrating a threshold for these parameters is available. One theory proposes that high levels of NaSac, combined with protein in a high Na+, high pH environment found only in the male rat, form toxic microscopic crystals; therefore, NaSac-induced tumors may not be relevant to human carcinogenesis.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Saccharin, ≥99%
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Saccharin, European Pharmacopoeia (EP) Reference Standard
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard