Merck
  • Rare earth (cerium oxide) pneumoconiosis: analytical scanning electron microscopy and literature review.
Rare earth (cerium oxide) pneumoconiosis: analytical scanning electron microscopy and literature review.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (1995-10-01)
J W McDonald, A J Ghio, C E Sheehan, P F Bernhardt, V L Roggli
ABSTRACT
Rare earth pneumoconiosis is an uncommonly reported disease caused by the inhalation of dust containing lanthanides, also known as rare earth metals, which are common industrial materials. The pathologic manifestations and natural history of this disorder are incompletely understood. We describe a male patient with a 35-year history of optical lens grinding, an occupation associated with exposure to cerium oxide, a rare earth metal-containing compound. The patient presented with progressive dyspnea and an interstitial pattern on chest X-ray; open lung biopsy showed interstitial fibrosis histologically indistinguishable from usual interstitial pneumonitis. However, scanning electron microscopy with energy-dispersive X-ray analysis demonstrated numerous particulate deposits in the lung, of which the majority contained the rare earth metal cerium alone or in combination with other elements. Our case is one of the first to describe rare earth pneumoconiosis associated with pulmonary fibrosis in the occupational setting of optical lens manufacture. Besides reinforcing the contention that rare earth metals are potentially harmful, our case suggests that such agents may be causally related to the development of pulmonary fibrosis.
Product Description
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis