Merck
  • Review: biocatalytic transformations of ferulic acid: an abundant aromatic natural product.
Review: biocatalytic transformations of ferulic acid: an abundant aromatic natural product.
Journal of industrial microbiology (1995-12-01)
J P Rosazza, Z Huang, L Dostal, T Volm, B Rousseau
ABSTRACT
In this review we examine the fascinating array of microbial and enzymatic transformations of ferulic acid. Ferulic acid is an extremely abundant, preformed phenolic aromatic chemical found widely in nature. Ferulic acid is viewed as a commodity scale, renewable chemical feedstock for biocatalytic conversion to other useful aromatic chemicals. Most attention is focused on bioconversions of ferulic acid itself. Topics covered include cinnamoyl side-chain cleavage; nonoxidative decarboxylation; mechanistic details of styrene formation; purification and characterization of ferulic acid decarboxylase; conversion of ferulic acid to vanillin; O-demethylation; and reduction reactions. Biotransformations of vinylguaiacol are discussed, and selected biotransformations of vanillic acid including oxidative and nonoxidative decarboxylation are surveyed. Finally, enzymatic oxidative dimerization and polymerization reactions are reviewed.
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
trans-Ferulic acid, 99%
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)