Merck
  • Platinum rechallenge in patients with advanced NSCLC: a pooled analysis.
Platinum rechallenge in patients with advanced NSCLC: a pooled analysis.
Lung cancer (Amsterdam, Netherlands) (2013-07-31)
Fausto Petrelli, Andrea Coinu, Mary Cabiddu, Mara Ghilardi, Mara Ardine, Sandro Barni
ABSTRACT
The sole agents pemetrexed (PEM), docetaxel and anti-EGFR agents are approved second-line therapies for non-small cell lung cancer (NSCLC) after failure with cisplatin-based doublets. The potential usefulness of platinum-based doublets as rechallenge for second-line chemotherapy has not yet been established. Studies that enrolled NSCLC platinum pre-treated patients were identified using electronic databases (MEDLINE and EMBASE). Pemetrexed and taxanes (TAXs)-based platinum combinations were considered. A systematic review was conducted using Comprehensive Meta-Analysis (version 2.2.064) software to calculate the event rate of response and 95% confidence interval. Median weighted progression-free survival (PFS) and overall survival (OS) time for PEM and TAXs-based doublets were compared by two-sided Student's t test. We tested for significant heterogeneity by Cochran's chi-square test and I(2) index. Eleven studies published between 1999 and 2012 were included in this analysis with a total of 607 patients enrolled; 468 were treated with PEM-doublets and 139 with TAXs-doublets. The overall response rate was 27.5% with a higher response rate of 37.8% (range, 29.7-46.7%) for TAXs-based treatment vs. 22% (range, 13.4-34.1%) for PEM-based combinations; (p < 0.0001). Median PFS and OS were 3.9 and 8.7 months with weighted PFS of 3.9 vs. 5.3 months (p < 0.0001) and similar OS for PEM vs. TAXs-based doublets. With the limitations of small and not randomised trials included, this pooled analysis shows that NSCLC patients who relapsed after a first-line platinum-based chemotherapy obtain a tumour response of 27% from a platinum rechallenge containing PEM or TAXs. Response rate and median PFS appear better with TAXs-than with PEM-doublets.
Sigma-Aldrich
Platinum, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, powder, 99.995% trace metals basis
Sigma-Aldrich
Platinum, powder (coarse), 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, thermocouple grade
Sigma-Aldrich
Platinum, foil, thickness 0.125-0.135 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, sponge, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.10 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.20 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum, nanopowder, <50 nm particle size (TEM)
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Platinum, wire reel, 0.1m, diameter 0.005mm, wollaston, 99.9%
Platinum, foil, 0.05m coil, thickness 0.125mm, coil width 2.5mm, as rolled, 99.99%
Platinum, foil, 0.2m coil, thickness 0.125mm, coil width 2.5mm, as rolled, 99.99%
Platinum, tube, 50mm, outside diameter 0.40mm, inside diameter 0.16mm, wall thickness 0.12mm, as drawn, 99.95%
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width 1.2mm, as rolled, 99.95%
Platinum, foil, 0.05m coil, thickness 0.15mm, coil width .9mm, as rolled, 99.99%
Platinum, foil, 0.5m coil, thickness 0.030mm, coil width .7mm, as rolled, 99.95%