Merck
Targeting and delivery of platinum-based anticancer drugs.
Chemical Society reviews (2012-10-09)
Xiaoyong Wang, Zijian Guo
ABSTRACT
Platinum-based anticancer drugs occupy a crucial role in the treatment of various malignant tumours. However, the efficacy and applicability of platinum drugs are heavily restricted by severe systemic toxicities and drug resistance. Different drug targeting and delivery (DTD) strategies have been developed to prevent the shortcomings of platinum-based chemotherapy. These approaches can be roughly categorized into two groups; namely, active and passive tactics. Active DTD is realized through specific molecular interactions between the drugs and cell or tissue elements, while passive DTD is achieved by exploiting the enhanced permeability and retention effect in tumour tissues. The principal methods for active DTD include conjugation of platinum drugs with selective targeting moieties or encapsulation of platinum drugs in host molecules. Bioactive substances such as hormones, carbohydrates, bisphosphonates, peptides and proteins are commonly used in active DTD. Passive DTD generally involves the fabrication of functionalized polymers or nanoparticles and the subsequent conjugation of platinum drugs with such entities. Polymeric micelles, liposomes, nanotubes and nanoparticles are frequently used in passive DTD. In some cases, both active and passive mechanisms are involved in one DTD system. This review concentrates on various targeting and delivery techniques for improving the efficacy and reducing the side effects of platinum-based anticancer drugs. The content covers most of the related literatures published since 2006. These innovative tactics represent current state-of-the-art developments in platinum-based anticancer drugs.
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum black, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Platinum, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99.95%
Pricing and availability is not currently available.
Platinum, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Pricing and availability is not currently available.
Platinum, tube, 200mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%
Pricing and availability is not currently available.
Platinum, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99.95%
Pricing and availability is not currently available.
Platinum, tube, 100mm, outside diameter 3.0mm, inside diameter 2.8mm, wall thickness 0.1mm, as drawn, 99.95%
Pricing and availability is not currently available.
Platinum, insulated wire, 1m, conductor diameter 0.025mm, insulation thickness 0.001mm, PTFE (polytetrafluoroethylene) insulation, 99.999%
Pricing and availability is not currently available.
Platinum, insulated wire, 2m, conductor diameter 0.05mm, insulation thickness 0.0075mm, polyester insulation, 99.99%
Pricing and availability is not currently available.
Platinum, rod, 25mm, diameter 1.0mm, 99.95%
Pricing and availability is not currently available.
Platinum, insulated wire, 0.5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyimide insulation, 99.99%
Pricing and availability is not currently available.
Platinum, rod, 200mm, diameter 1.0mm, 99.95%
Pricing and availability is not currently available.
Platinum, foil, 30x30mm, thickness 0.2mm, as rolled, 99.95%
Pricing and availability is not currently available.
Platinum, tube, 100mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%
Pricing and availability is not currently available.
Platinum, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.009mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Pricing and availability is not currently available.
Platinum, wire reel, 0.05m, diameter 0.8mm, annealed, may be used as one component of a thermocouple, 99.99%
Pricing and availability is not currently available.
Platinum, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.0075mm, polyester insulation, 99.99%
Pricing and availability is not currently available.
Platinum, tube, 50mm, outside diameter 3.0mm, inside diameter 2.8mm, wall thickness 0.1mm, as drawn, 99.95%
Pricing and availability is not currently available.
Platinum, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Pricing and availability is not currently available.