Merck
  • HLA genotype and carbamazepine-induced cutaneous adverse drug reactions: a systematic review.
HLA genotype and carbamazepine-induced cutaneous adverse drug reactions: a systematic review.
Clinical pharmacology and therapeutics (2012-11-08)
V L Yip, A G Marson, A L Jorgensen, M Pirmohamed, A Alfirevic
ABSTRACT
Carbamazepine (CBZ) therapy is associated with cutaneous adverse reactions in up to 10% of patients. Predisposition to these hypersensitivity reactions has been linked to the human leukocyte antigen (HLA) genotype. This systematic review determines the strength of these associations and accuracy of proposed genetic screening. We determined that carriage of HLA-B*1502 in Asian patients was associated with a pooled odds ratio (OR) of 113.4 (95% confidence interval (CI) = 51.2-251.0, P < 1 × 10(-5)) for CBZ-induced Stevens-Johnson syndrome (SJS) and toxic epidermal necrolysis (TEN). A total of 461 patients would need to be screened for HLA-B*1502 to prevent one episode of SJS/TEN. HLA-A*3101 is significantly associated with all phenotypes of CBZ hypersensitivity in multiple ethnicities with a pooled OR of 9.5 (95% CI = 6.4-13.9, P < 1 × 10(-5)). Between 47 and 67 patients would need to be tested for HLA-A*3101 to prevent one episode of hypersensitivity. Our findings suggest that HLA testing before carbamazepine therapy would be effective at identifying individuals at risk of hypersensitivity and applicable to multiple populations providing hope for prevention in the future.
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, analytical standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®