Merck
Fluconazole pharmacokinetics and safety in premature infants.
Current medicinal chemistry (2012-08-11)
K Turner, P Manzoni, D K Benjamin, M Cohen-Wolkowiez, P B Smith, M M Laughon
ABSTRACT
Invasive candidiasis (IC) in the premature infant population is a common infection that results in substantial morbidity and mortality. For these patients, fluconazole is among the first line therapies to treat and prevent IC, and yet few prospective studies investigating its pharmacokinetics (PK) and safety have been performed in this vulnerable population. We review five phase I studies examining the PK of fluconazole in premature infants, which demonstrate markedly differing kinetics compared to adults. Based on these data, a treatment dose of 12 mg/kg/day, with the potential need of a loading dose of 25 mg/kg to achieve rapid steady state concentrations, achieves surrogate pharmacodynamic targets. Additionally, fluconazole appears to be safe to use in this population, with only minimal reversible hepatobiliary effects.
Product Description
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluconazole solution, 2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard