Merck
Molybdenum enzymes and molybdenum cofactor in mycobacteria.
Journal of cellular biochemistry (2011-06-17)
Tingyu Shi, Jianping Xie
ABSTRACT
When intracelluar pathogens enter the host macrophages where in addition to oxidative and antibiotic mechanisms of antimicrobial activity, nutrients are deprived. Human pathogen Mycobacterium tuberculosis is one of macrophage parasitisms, which can replicate and persist for decades in dormancy state in virulent environments. It is very successful in escaping the killing mechanisms of macrophage. Molybdenum (Mo) enzymes involve in the global carbon, sulfur, and nitrogen cycles by catalyzing important redox reactions. There are several Mo enzymes in mycobacteria and they exert several important physiological functions, such as dormancy regulation, the metabolism of energy sources, and nitrogen source. Pterin-based Mo cofactor (Moco) is the common cofactor of the Mo enzymes in mycobacteria but the cofactor biosynthesis is nearly an untapped area. The present article discusses the physiological function of Mo enzymes and the structural feature of the genes coding for Moco biosynthesis enzymes in mycobacteria.
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Molybdenum, foil, 6mm disks, thickness 0.03mm, annealed, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.05mm, annealed, 99.9%
Molybdenum, rod, 100mm, diameter 25mm, centerless ground, 99.9%
Molybdenum, rod, 200mm, diameter 5.0mm, centerless ground, 99.9%
Molybdenum, foil, light tested, 29x200mm, thickness 0.0712mm, annealed, 99.9%
Molybdenum, rod, 1000mm, diameter 8.0mm, centerless ground, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.009mm, 99.9%
Molybdenum, foil, not light tested, 150x150mm, thickness 0.05mm, annealed, 99.9%
Molybdenum, foil, 100x100mm, thickness 0.25mm, annealed, polished on both sides, 99.9%
Molybdenum, foil, not light tested, 100x100mm, thickness 0.005mm, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.004mm, 99.9%
Molybdenum, microfoil, disks, 25mm, thinness 1.0μm, specific density 1022μg/cm2, permanent mylar 3.5μm support, 99.9%
Molybdenum, wire reel, 100m, diameter 0.03mm, hard, 99.95%
Molybdenum, wire reel, 100m, diameter 0.013mm, hard, 99.8+%
Molybdenum, foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.9%
Molybdenum, mesh, 150x150mm, nominal aperture 0.44mm, wire diameter 0.07mm, 50 wires/inch, open area 67%, plain weave mesh
Molybdenum, foil, 150x150mm, thickness 0.125mm, annealed, 99.9%
Molybdenum, foil, 100x100mm, thickness 0.5mm, polished on both sides, 99.9%
Molybdenum, wire reel, 10m, diameter 0.013mm, hard, 99.8+%
Molybdenum, wire reel, 1m, diameter 0.5mm, annealed, 99.95%
Molybdenum, pellets, 50g, max. size 10mm, 99.9%
Molybdenum, wire reel, 100m, diameter 0.025mm, gold plated, 99.95%