Merck
Pharmacological potential of cerium oxide nanoparticles.
Nanoscale (2011-03-04)
Ivana Celardo, Jens Z Pedersen, Enrico Traversa, Lina Ghibelli
ABSTRACT
Nanotechnology promises a revolution in pharmacology to improve or create ex novo therapies. Cerium oxide nanoparticles (nanoceria), well-known as catalysts, possess an astonishing pharmacological potential due to their antioxidant properties, deriving from a fraction of Ce(3+) ions present in CeO(2). These defects, compensated by oxygen vacancies, are enriched at the surface and therefore in nanosized particles. Reactions involving redox cycles between the Ce(3+) and Ce(4+) oxidation states allow nanoceria to react catalytically with superoxide and hydrogen peroxide, mimicking the behavior of two key antioxidant enzymes, superoxide dismutase and catalase, potentially abating all noxious intracellular reactive oxygen species (ROS) via a self-regenerating mechanism. Hence nanoceria, apparently well tolerated by the organism, might fight chronic inflammation and the pathologies associated with oxidative stress, which include cancer and neurodegeneration. Here we review the biological effects of nanoceria as they emerge from in vitro and in vivo studies, considering biocompatibility and the peculiar antioxidant mechanisms.
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium, chips
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Cerium, foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%