Merck
  • Assessment of the environmental toxicity and carcinogenicity of tungsten-based shot.
Assessment of the environmental toxicity and carcinogenicity of tungsten-based shot.
Ecotoxicology and environmental safety (2009-02-24)
Vernon G Thomas, Michael J Roberts, Paul T C Harrison
ABSTRACT
The toxicity of elemental tungsten released from discharged shot was assessed against previous studies that established a 1% toxic threshold for soil organisms. Extremely heavy theoretical shot loadings of 69,000shot/ha were used to generate estimated environmental concentrations (EEC) for two brands of tungsten-based shot containing 51% and 95% tungsten. The corresponding tungsten EEC values were 6.5-13.5mg W/kg soil, far below the 1% toxic threshold. The same shot loading in water produced tungsten EEC values of 2.1-4.4mg W/L, levels that are not toxic under experimental conditions. Pure tungsten has not been shown to exhibit carcinogenic properties when ingested or embedded in animal tissues, but nickel, with which it is often alloyed, has known carcinogenicity. Given the large number of waterfowl that carry shot embedded in their body, it is advisable to screen lead shot substitutes for their carcinogenic potential through intra-muscular implantation.
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Tungsten, insulated wire, 5m, conductor diameter 0.075mm, insulation thickness 0.02mm, PTFE (polytetrafluoroethylene) insulation, 99.9%
Tungsten, insulated wire, 10m, conductor diameter 0.05mm, insulation thickness 0.008mm, polyimide insulation, 99.9%
Tungsten, microfoil, disks, 10mm, thinness 0.25μm, specific density 474μg/cm2, permanent mylar 3.5μm support, 99.95%
Tungsten, insulated wire, 0.2m, conductor diameter 0.015mm, insulation thickness 0.002mm, polyimide insulation, 99.9%
Tungsten, foil, 5m coil, thickness 0.025mm, coil width 1mm, as rolled, 99.95%
Tungsten, insulated wire, 20m, conductor diameter 0.05mm, insulation thickness 0.008mm, polyimide insulation, 99.9%
Tungsten, foil, not light tested, 50x50mm, thickness 0.004mm, 99.95%
Tungsten, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95%
Tungsten, lump, 200g, max. size 5mm
Tungsten, insulated wire, 2m, conductor diameter 0.075mm, insulation thickness 0.02mm, PTFE (polytetrafluoroethylene) insulation, 99.9%
Tungsten, rod, 200mm, diameter 6.35mm, centerless ground, 99.95%
Tungsten, foil, light tested, 50x50mm, thickness 0.075mm, as rolled, 99.95%
Tungsten, foil, not light tested, 25x25mm, thickness 0.004mm, 99.95%
Tungsten, insulated wire, 2m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyimide insulation, 99.9%
Tungsten, foil, 30mm disks, thickness 1.0mm, as rolled, 99.95%
Tungsten, foil, light tested, 150x150mm, thickness 0.075mm, as rolled, 99.95%
Tungsten, lump, 50g, max. size 5mm
Tungsten, insulated wire, 1m, conductor diameter 0.015mm, insulation thickness 0.002mm, polyimide insulation, 99.9%
Tungsten, microfoil, disks, 10mm, thinness 0.05μm, specific density 96μg/cm2, permanent mylar 3.5μm support, 99.95%
Tungsten, insulated wire, 2m, conductor diameter 0.0125mm, insulation thickness 0.0035mm, polyimide insulation, 99.9%
Tungsten, foil, light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95%
Tungsten, microfoil, disks, 10mm, thinness 0.1μm, specific density 182μg/cm2, permanent mylar 3.5μm support, 99.95%
Tungsten, wire reel, 0.5m, diameter 1.0mm, clean, 99.95%
Tungsten, insulated wire, 2m, conductor diameter 0.015mm, insulation thickness 0.002mm, polyimide insulation, 99.9%
Tungsten, foil, not light tested, 50x50mm, thickness 0.007mm, 99.95%
Tungsten, foil, not light tested, 25x25mm, thickness 0.006mm, 99.95%
Tungsten, rod, 100mm, diameter 5.0mm, centerless ground, 99.95%
Tungsten, mesh, 100x100mm, nominal aperture 0.2mm, wire diameter 0.05mm, 100 wires/inch, open area 64%, plain weave mesh, 99.95%