Merck
  • Mebendazole in parasitic infections other than those caused by soil-transmitted helminths.
Mebendazole in parasitic infections other than those caused by soil-transmitted helminths.
Transactions of the Royal Society of Tropical Medicine and Hygiene (2009-02-07)
R Cañete, A A Escobedo, P Almirall, M E González, K Brito, S Cimerman
ABSTRACT
Mebendazole, a benzimidazole carbamate compound, is currently in use for human medical practice against soil-transmitted helminthiasis (STH) and enterobiasis. However, it has been demonstrated that its spectrum of activity is broad and goes beyond those infections. Several studies provide evidence that this drug, taken at higher doses than used for STH and enterobiasis, could be sufficiently effective on some protozoa, nematodes and cestodes.
Product Description
Mebendazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Mebendazole, VETRANAL®, analytical standard
Supelco
Mebendazole, analytical standard, ≥98% (HPLC)
USP
Mebendazole, United States Pharmacopeia (USP) Reference Standard
Mebendazole for system suitability, European Pharmacopoeia (EP) Reference Standard