Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Pricing and availability is not currently available.
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 5m, diameter 0.25mm, hard, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
Pricing and availability is not currently available.
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Pricing and availability is not currently available.
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Pricing and availability is not currently available.
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Pricing and availability is not currently available.
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Pricing and availability is not currently available.
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Pricing and availability is not currently available.
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Pricing and availability is not currently available.
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Pricing and availability is not currently available.
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Pricing and availability is not currently available.