Merck
  • Nasal allergies and beyond: a clinical review of the pharmacology, efficacy, and safety of mometasone furoate.
Nasal allergies and beyond: a clinical review of the pharmacology, efficacy, and safety of mometasone furoate.
Allergy (2005-06-14)
C van Drunen, E O Meltzer, C Bachert, J Bousquet, W J Fokkens
ABSTRACT
Mometasone furoate nasal spray (MFNS; Nasonex, Schering-Plough Corporation, Kenilworth, NJ, USA) is an effective and well-tolerated intranasal corticosteroid approved for the prophylactic treatment of seasonal allergic rhinitis, and the treatment of perennial allergic rhinitis. MFNS is a potent molecule with a rapid onset of action and excellent safety and efficacy profiles. Having recently received approval for the treatment of nasal polyposis, data indicate that MFNS may also be effective in rhinosinusitis.
Product Description
Mometasone furoate, European Pharmacopoeia (EP) Reference Standard
USP
Mometasone furoate, United States Pharmacopeia (USP) Reference Standard
Supelco
Mometasone Furoate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mometasone furoate, ≥98% (HPLC)