- A review of technology and safety aspects of erbium lasers in dentistry.
A review of technology and safety aspects of erbium lasers in dentistry.
Dental update (2001-08-31)
D M Clarkson
PMID11526883
ABSTRACT
This article reviews aspects of the probable mechanisms used by erbium dental lasers for cutting dentine and enamel, describes key issues of the risk of temperature elevation and speed of cutting relative to conventional techniques and looks at issues concerned with the safety of lasers.
Product Description
Erbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Erbium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Erbium, foil, 25x25mm, thickness 1.0mm, as rolled, 99%
Erbium, rod, 50mm, diameter 2.0mm, cast, 99%
Erbium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Erbium, rod, 50mm, diameter 10.0mm, cast, 99%
Erbium, rod, 50mm, diameter 6.35mm, cast, 99%
Erbium, rod, 100mm, diameter 10.0mm, cast, 99%
Erbium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Erbium, rod, 100mm, diameter 6.35mm, cast, 99%
Erbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Erbium, foil, 50x50mm, thickness 1.0mm, as rolled, 99%
Erbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%