Merck
  • Geochemical associations and availability of cadmium (Cd) in a paddy field system, northwestern Thailand.
Geochemical associations and availability of cadmium (Cd) in a paddy field system, northwestern Thailand.
Environmental pollution (Barking, Essex : 1987) (2014-02-08)
Peerapat Kosolsaksakul, John G Farmer, Ian W Oliver, Margaret C Graham
ABSTRACT
The Mae Tao watershed, northwest Thailand, has become contaminated with cadmium (Cd) as a result of zinc ore extraction (Padaeng deposit) in the nearby Thanon-Thongchai mountains. Consumption of contaminated rice has led to documented human health impacts. The aim of this study was to elucidate transfer pathways from creek and canal waters to the paddy field soils near Baan Mae Tao Mai village and to determine the relationship between Cd speciation in the soil and uptake by rice plants. Transfer mainly occurred in association with particulate matter during flooding and channel dredging and, in contrast with many other studies, most of the soil Cd was associated with exchangeable and carbonate-bound fractions. Moreover, there was a linear relationship between soil total Cd and rice grain Cd (R(2) = 0.715), but a stronger relationship between both the Tessier-exchangeable soil Cd and the BCR-exchangeable soil Cd and rice grain Cd (R(2) = 0.898 and 0.862, respectively).
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, wire reel, 0.1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, wire reel, 0.1m, diameter 2.0mm, hard, 99.99+%
Cadmium, wire reel, 0.1m, diameter 0.70mm, as drawn, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, rod, 150mm, diameter 10.0mm, as drawn, 99.99+%
Cadmium, tube, 200mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, wire reel, 10m, diameter 0.25mm, hard, 99.99+%
Cadmium, foil, 10mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, 30x100mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 99.95%
Cadmium, wire reel, 0.2m, diameter 0.70mm, as drawn, 99.99+%
Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 10mm disks, thickness 0.025mm, 99.7+%
Cadmium, foil, 30x150mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Cadmium, wire reel, 0.5m, diameter 2.0mm, hard, 99.99+%
Cadmium, foil, 50x50mm, thickness 2.0mm, as rolled, 99.99%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%