Merck
  • Dietary nitrate and nitrite intake and risk of colorectal cancer in the Shanghai Women's Health Study.
Dietary nitrate and nitrite intake and risk of colorectal cancer in the Shanghai Women's Health Study.
International journal of cancer (2013-11-19)
Curt T Dellavalle, Qian Xiao, Gong Yang, Xiao-Ou Shu, Briseis Aschebrook-Kilfoy, Wei Zheng, Hong Lan Li, Bu-Tian Ji, Nathaniel Rothman, Wong-Ho Chow, Yu-Tang Gao, Mary H Ward
ABSTRACT
Nitrate and nitrite are precursors of endogenously formed N-nitroso compounds (NOC), known animal carcinogens. Nitrosation reactions forming NOCs can be inhibited by vitamin C and other antioxidants. We prospectively investigated the association between dietary nitrate and nitrite intake and risk of colorectal cancer in the Shanghai Women's Health Study, a cohort of 73,118 women ages 40-70 residing in Shanghai. We evaluated effect modification by factors that affect endogenous formation of NOCs: vitamin C (at or above/below median) and red meat intake (at or above/below median). Nitrate, nitrite and other dietary intakes were estimated from a 77-item food frequency questionnaire administered at baseline. Over a mean of 11 years of follow-up, we identified 619 colorectal cancer cases (n = 383, colon; n = 236, rectum). Hazard ratios (HR) and 95% confidence intervals (CI) were estimated using Cox proportional hazard regression. Overall, nitrate intake was not associated with colorectal cancer risk (HR = 1.08; 95% CI: 0.73-1.59). However, among women with vitamin C intake below the median (83.9 mg day(-1) ) and hence higher potential exposure to NOCs, risk of colorectal cancer increased with increasing quintiles of nitrate intake (highest vs. lowest quintile HR = 2.45; 95% CI: 1.15-5.18; p trend = 0.02). There was no association among women with higher vitamin C intake. We found no association between nitrite intake and risk of colorectal cancer overall or by intake level of vitamin C. Our findings suggest that high dietary nitrate intake among subgroups expected to have higher exposure to endogenously formed NOCs increases risk of colorectal cancer.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard