Sigma-Aldrich
Atto 390 azide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 840 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 100 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 820 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 6 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 840 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 18 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 1000 nm (IR-II), maleimide-thiol functionalized, 1 ea of 60 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 510B nm (green), carboxy-NHS-amine functionalized, 1 ea of 60 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 510B nm (green), carboxy-NHS-amine functionalized, 1 ea of 6 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 475 nm (blue), carboxy-NHS-amine functionalized, 1 ea of 100 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 435 nm (indigo), maleimide-thiol functionalized, 1 ea of 60 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 550 nm (yellow), carboxy-NHS-amine functionalized, 1 ea of 18 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 840 nm (IR-I), maleimide-thiol functionalized, 1 ea of 6 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 770 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 100 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 660 nm (red), carboxy-NHS-amine functionalized, 1 ea of 6 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 435 nm (indigo), carboxy-NHS-amine functionalized, 1 ea of 10 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 840 nm (IR-I), maleimide-thiol functionalized, 1 ea of 30 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 770 nm (IR-I), maleimide-thiol functionalized, 1 ea of 10 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 420 nm (violet), carboxy-NHS-amine functionalized, 1 ea of 60 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 840 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 6 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 770 nm (IR-I), maleimide-thiol functionalized, 1 ea of 60 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 610 nm (orange), maleimide-thiol functionalized, 1 ea of 18 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 530 nm (green), maleimide-thiol functionalized, 1 ea of 6 nmol oligonucleotide kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 610 nm (orange), maleimide-thiol functionalized, 1 ea of 100 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 580 nm (yellow), maleimide-thiol functionalized, 1 ea of 100 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 830 nm (IR-I), maleimide-thiol functionalized, 1 ea of 10 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 580 nm (yellow), maleimide-thiol functionalized, 1 ea of 30 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 820 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 30 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 660 nm (red), carboxy-NHS-amine functionalized, 1 ea of 10 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 550 nm (yellow), maleimide-thiol functionalized, 1 ea of 100 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 510 nm (green), maleimide-thiol functionalized, 1 ea of 30 μg antibody kit
Sigma-Aldrich
Conjugated Polymer Nanoparticle (CPN), fluorescence λem 840 nm (IR-I), maleimide-thiol functionalized, 1 ea of 10 μg antibody kit