Merck
  • Final overall survival: fulvestrant 500 mg vs 250 mg in the randomized CONFIRM trial.
Final overall survival: fulvestrant 500 mg vs 250 mg in the randomized CONFIRM trial.
Journal of the National Cancer Institute (2013-12-10)
Angelo Di Leo, Guy Jerusalem, Lubos Petruzelka, Roberto Torres, Igor N Bondarenko, Rustem Khasanov, Didier Verhoeven, José L Pedrini, Iya Smirnova, Mikhail R Lichinitser, Kelly Pendergrass, Luca Malorni, Sally Garnett, Yuri Rukazenkov, Miguel Martin
ABSTRACT
At the time of the initial analysis of overall survival (OS) for the Comparison of Faslodex in Recurrent or Metastatic Breast Cancer (CONFIRM) randomized, double-blind, phase III trial, approximately 50% of patients had died. A final analysis of OS was subsequently planned for when 75% of patients had died. Patients were randomly assigned 1:1 to fulvestrant 500 mg administered as two 5-mL intramuscular injections on days 0, 14, and 28 and every 28 (±3) days thereafter or fulvestrant 250 mg administered as two 5-mL intramuscular injections (one fulvestrant and one placebo [identical in appearance to study drug]) on days 0, 14 (two placebo injections only), and 28 and every 28 (±3) days thereafter. OS was analyzed using an unadjusted log-rank test. No adjustments were made for multiplicity. Serious adverse events (SAEs) and best response to subsequent therapy were also reported. All statistical tests were two-sided. In total, 736 women (median age = 61.0 years) were randomly assigned to fulvestrant 500 mg (n = 362) or 250 mg (n = 374). At the final survival analysis, 554 of 736 (75.3%) patients had died. Median OS was 26.4 months for fulvestrant 500 mg and 22.3 months for 250 mg (hazard ratio = 0.81; 95% confidence interval = 0.69-0.96; nominal P = .02). There were no clinically important differences in SAE profiles between the treatment groups; no clustering of SAEs could be detected in either treatment group. Type of first subsequent therapy and objective responses to first subsequent therapy were well balanced between the two treatment groups. In patients with locally advanced or metastatic estrogen receptor-positive breast cancer, fulvestrant 500 mg is associated with a 19% reduction in risk of death and a 4.1-month difference in median OS compared with fulvestrant 250 mg. Fulvestrant 500 mg was well tolerated, and no new safety concerns were identified.
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications