Merck
  • Elevated plasma vitamin B12 levels as a marker for cancer: a population-based cohort study.
Elevated plasma vitamin B12 levels as a marker for cancer: a population-based cohort study.
Journal of the National Cancer Institute (2013-11-20)
Johan Frederik Berg Arendt, Lars Pedersen, Ebba Nexo, Henrik Toft Sørensen
ABSTRACT
A substantial proportion of patients referred for plasma vitamin B12 (cobalamin [Cbl]) measurement present with high Cbl levels, which have been reported in patients with different cancer types. However, the cancer risk among patients with newly diagnosed high Cbl levels has not been adequately examined. We conducted this cohort study using population-based Danish medical registries. Patients referred for Cbl measurement with levels greater than the lower reference limit (≥200 pmol/L) were identified from the population of Northern Denmark during the period of 1998 to 2009 using a database of laboratory test results covering the entire population. Data on cancer incidence (follow-up 1998-2010), Cbl treatment, and prior diagnoses were obtained from medical registries. Patients receiving Cbl treatment were excluded. Cancer risks were calculated as standardized incidence ratios (SIRs) with 95% confidence intervals (CIs), stratified by plasma Cbl levels. All statistical tests were two-sided. We identified 333 667 persons without prevalent cancer and not receiving Cbl treatment. Six percent had Cbl levels greater than the upper reference limit (≥601 pmol/L). Cancer risk increased with higher Cbl levels and was highest during the first year of follow-up (Cbl 601-800 pmol/L: SIR = 3.44, 95% CI = 3.14 to 3.76; Cbl >800 pmol/L: SIR = 6.27, 95% CI = 5.70 to 6.88; both P < .001). The risks were particularly elevated for hematological and smoking- and alcohol-related cancers for persons with high Cbl levels. High Cbl levels were associated with the risk of subsequently diagnosed cancer, mostly within the first year of follow-up. This may have clinical implications for the interpretation of high Cbl levels.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Supelco
Cyanocobalamin (B12), analytical standard
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material