Merck
  • Amplification of steroid-mediated SP-B expression by physiological levels of caffeine.
Amplification of steroid-mediated SP-B expression by physiological levels of caffeine.
American journal of physiology. Lung cellular and molecular physiology (2013-10-29)
Markus Fehrholz, Matthias Hütten, Boris W Kramer, Christian P Speer, Steffen Kunzmann
ABSTRACT
Factors positively influencing surfactant homeostasis in general and surfactant protein B (SP-B) expression in particular are considered of clinical importance regarding an improvement of lung function in preterm infants. The objective of this study was to identify effects of physiological levels of caffeine on glucocorticoid-mediated SP-B expression in vitro and in vivo. Levels of SP-B and pepsinogen C were quantified by quantitative real-time RT-PCR or immunoblotting in NCI-H441 cells daily exposed to caffeine and/or dexamethasone (DEX). In vivo, SP-B expression was analyzed in bronchoalveolar lavage (BAL) of preterm sheep exposed to antenatal DEX and/or postnatal caffeine. If DEX and caffeine were continuously present, SP-B mRNA and protein levels were increased for up to 6 days after induction (P < 0.05). Additionally, caffeine enhanced SP-B mRNA expression in DEX-pretreated cells (P < 0.05). Moreover, caffeine amplified DEX-induced pepsinogen C mRNA expression (P < 0.05). After short-term treatment with caffeine in vivo, only slightly higher SP-B levels could be detected in BAL of preterm sheep following antenatal DEX, combined with an increase of arterial oxygen partial pressure (P < 0.01). Our data demonstrated that the continuous presence of caffeine in vitro is able to amplify DEX-mediated SP-B expression. In contrast, short-term improvement of lung function in vivo is likely to be independent of altered SP-B transcription and translation. An impact of caffeine on release of surfactant reservoirs from lamellar bodies could, however, quickly affect SP-B content in BAL, which has to be further investigated. Our findings indicate that caffeine is able to amplify main effects of glucocorticoids that result from changes in surfactant production, maturation, and release.
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Caffeine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard