Merck
  • Zinc finger proteins: new insights into structural and functional diversity.
Zinc finger proteins: new insights into structural and functional diversity.
Current opinion in structural biology (2001-02-17)
J H Laity, B M Lee, P E Wright
ABSTRACT
Zinc finger proteins are among the most abundant proteins in eukaryotic genomes. Their functions are extraordinarily diverse and include DNA recognition, RNA packaging, transcriptional activation, regulation of apoptosis, protein folding and assembly, and lipid binding. Zinc finger structures are as diverse as their functions. Structures have recently been reported for many new zinc finger domains with novel topologies, providing important insights into structure/function relationships. In addition, new structural studies of proteins containing the classical Cys(2)His(2) zinc finger motif have led to novel insights into mechanisms of DNA binding and to a better understanding of their broader functions in transcriptional regulation.
Sigma-Aldrich
Anti-ZNF121 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ZNF607 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ZNF83 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ZNF79 antibody produced in rabbit, affinity isolated antibody