Merck
  • Structural and electronic factors in heterolytic cleavage: formation of the Co(I) intermediate in the corrinoid/iron-sulfur protein from Clostridium thermoaceticum.
Structural and electronic factors in heterolytic cleavage: formation of the Co(I) intermediate in the corrinoid/iron-sulfur protein from Clostridium thermoaceticum.
Biochemistry (1995-04-18)
M D Wirt, M Kumar, J J Wu, E M Scheuring, S W Ragsdale, M R Chance
ABSTRACT
We have completed the first direct structural characterization of an enzyme-bound four-coordinate Co(I) intermediate, in this case for the corrinoid/iron-sulfur protein (C/Fe-SP) from Clostridium thermoaceticum. Extended X-ray absorption fine structure and X-ray edge spectroscopy of the active Co(I) state of the C/Fe-SP indicates a four-coordinate (distorted) square-planar structure where the best fit gives average Co-N(equatorial) distances of 1.87 +/- 0.01 A, corresponding to 4.2 +/- 0.3 ligands. The X-ray edge spectrum of Co(I) C/Fe-SP contains a moderate intensity 1s-4p + "shake-down" (SD) transition and no 1s-3d peak (where SD transitions are indicative of square-planar geometries). X-ray edge results for the methyl-Co(III) form, reported earlier [Wirt, M. D., Kumar, M., Ragsdale, S. W., & Chance, M. R. (1993) J. Am. Chem. Soc. 115, 2146-2150], are consistent with a base-off methylcobamide structure. The absence of a ligated 5-methoxybenzimidazole base in the methyl-Co(III) state is important since the base-off form is predicted to predispose the Co-C bond toward heterolytic cleavage to form the four-coordinate Co(I) species concurrent with methyl transfer. Additionally, we have examined first-derivative X-ray edge spectra of Co(I) C/Fe-SP, relative to edge spectra of a cobalt foil, as an indicator of effective nuclear charge on cobalt. The Co(I) C/Fe-SP edge position at 7720.5 +/- 0.3 eV is less than, but very close to, the value seen for the corresponding free Co(I) cobalamin.(ABSTRACT TRUNCATED AT 250 WORDS)
Product Description
Cobalt, foil, 8mm disks, thickness 0.004mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.003mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.02mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 50mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 4mm disks, thickness 0.005mm, 99.9%
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.005mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.004mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 8mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 6mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, light tested, 50x50mm, thickness 0.015mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.9%
Cobalt, foil, 8mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 6mm disks, thickness 0.005mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.015mm, 99.9%