- A novel sample introduction technique for combustion total organic carbon analysis in aqueous materials.
A novel sample introduction technique for combustion total organic carbon analysis in aqueous materials.
Talanta (1994-10-01)
W K Winnett, M P Murphy
PMID18966112
ABSTRACT
A modification of the sample introduction system for combustion total organic carbon analyzers is presented where sample is sprayed into the instrument. Thermally created sprays and nebulizer sprays are discussed. It is shown that spray introduction gives more symmetric signals, less effective noise and less signal tailing than conventional drip sample introduction. This performance improvement can be especially important in applications where the aqueous sample contains dissolved solids and corrosive materials. Lining the inside of the combustion tube with nickel foil also provided an increase in the lifetime due to protection from thermal shock from the cooler liquid sample.
Product Description
Nickel, foil, 4mm disks, thickness 0.125mm, as rolled, 100%
Nickel, foil, 5m coil, thickness 0.025mm, annealed, 99%
Nickel, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 50mm disks, thickness 0.50mm, annealed, 99%
Nickel, foil, 50mm disks, thickness 0.03mm, 99.9%
Nickel, foil, 4mm disks, thickness 0.20mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.075mm, annealed, 99%
Nickel, foil, 5m coil, thickness 0.05mm, annealed, 99%
Nickel, foil, 50mm disks, thickness 0.25mm, annealed, 99%
Nickel, foil, 50mm disks, thickness 0.15mm, annealed, 99%
Nickel, foil, 50mm disks, thickness 0.0025mm, 99.95%
Nickel, foil, 50mm disks, thickness 0.0125mm, 99.99+%
Nickel, foil, 4mm disks, thickness 0.25mm, as rolled, 100%
Nickel, foil, 4mm disks, thickness 0.50mm, annealed, 99%
Nickel, foil, 5m coil, thickness 0.05mm, coil width 2mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.5mm, annealed, 99.98%
Nickel, foil, 50mm disks, thickness 0.01mm, 99.95%
Nickel, foil, 4mm disks, thickness 0.25mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.125mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.5mm, as rolled, 100%
Nickel, foil, 1m coil, thickness 0.004mm, hard, 99%
Nickel, foil, 10mm disks, thickness 0.05mm, as rolled, 100%
Nickel, foil, 10mm disks, thickness 0.1mm, annealed, 99%
Nickel, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Nickel, foil, 1m coil, thickness 0.075mm, as rolled, 99.98%
Nickel, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 10mm disks, thickness 0.125mm, as rolled, 100%
Nickel, foil, 1m coil, thickness 0.075mm, annealed, 99%
Nickel, foil, 1m coil, thickness 0.038mm, 99.9%
Nickel, foil, 10mm disks, thickness 0.20mm, annealed, 99%