Merck
  • Sequencing platinum foil matrix removal in postceramic soldering of the collarless veneered retainer.
Sequencing platinum foil matrix removal in postceramic soldering of the collarless veneered retainer.
The International journal of prosthodontics (1991-09-01)
S M Hannon, R B Gunderson, L Lorton, T Zislis, S O Hondrum
ABSTRACT
Postceramic soldering of collarless veneered retainers in which the porcelain margins were formed with a platinum foil matrix technique was investigated. The purpose of the study was to determine if removing the platinum foil matrix before or after indexing and soldering procedures would affect the porcelain margin integrity in fixed partial dentures. Prostheses were fabricated on a nickel-chromium laboratory model using both sequences. Six test cycles were performed. Each cycle included one soldering with matrix support and one without the matrix, for a total of 12 solderings. The fixed partial dentures were compared for degree of marginal seating with a measuring microscope and for configuration changes at the porcelain margins with scanning electron micrographs. In the microscopic analysis of marginal closure, soldering without foil matrix was statistically equal to soldering with foil in place. Removal of the platinum foil matrices prior to indexing provided for no further seating of the metal ceramic retainers. Evaluation by scanning electron microscopy demonstrated distinct configuration changes in the porcelain margins for the specimens soldered without matrix support.
Product Description
Platinum, foil, not light tested, 50x50mm, thickness 0.004mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99.95%
Platinum, foil, 30x30mm, thickness 0.2mm, as rolled, 99.95%
Platinum, foil, 15x15mm, thickness 0.2mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Platinum, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, not light tested, 50x50mm, thickness 0.0025mm, as rolled, 99.95%
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Platinum, foil, 0.1m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, 4mm disks, thickness 0.125mm, as rolled, 99.95%
Platinum, foil, 10mm disks, thickness 0.0125mm, as rolled, 99.95%
Platinum, foil, 4mm disks, thickness 0.05mm, as rolled, 99.95%
Platinum, foil, 15mm disks, thickness 0.01mm, as rolled, 99.95%
Platinum, foil, 6mm disks, thickness 0.025mm, as rolled, 99.99+%
Platinum, foil, 8mm disks, thickness 0.1mm, as rolled, 99.99+%
Platinum, foil, 4mm disks, thickness 0.004mm, as rolled, 99.95%
Platinum, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Platinum, foil, 8mm disks, thickness 0.0075mm, as rolled, 99.95%
Platinum, foil, 4mm disks, thickness 0.025mm, as rolled, 99.95%
Platinum, foil, 15mm disks, thickness 0.025mm, as rolled, 99.95%
Platinum, foil, light tested, 25x25mm, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, 0.5m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Platinum, foil, not light tested, 100x100mm, thickness 0.003mm, as rolled, 99.95%
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Platinum, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Platinum, foil, 6mm disks, thickness 0.004mm, as rolled, 99.95%
Platinum, foil, 15mm disks, thickness 0.025mm, as rolled, 99.99+%
Platinum, foil, 0.2m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%